Learn More
Barium nitrate, 99+%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 592.0°C |
| Color | White |
| pH | 5 to 8 (5% soln. at 20°C) |
| Physical Form | Crystals |
| Quantity | 2.5 kg |
| Assay Percent Range | 99% min. (Complexometry) |
| Linear Formula | Ba(NO3)2 |
| Merck Index | 15, 979 |
| Solubility Information | Solubility in water: 9g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone |
| IUPAC Name | barium(2+);dinitrate |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Harmful if inhaled. May intensify fire; oxidizer. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfor |
| EINECSNumber | 233-020-5 |
| RTECSNumber | CQ9625000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.