Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium nitrate, pure

Catalog No. AC453800100
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453800100 10 kg
AC453805000 500 g
AC453800025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC453800100 Supplier Thermo Scientific Chemicals Supplier No. 453800100
Add to Cart

CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 592.0°C
Quantity 10 kg
Linear Formula Ba(NO3)2
Merck Index 15,979
Solubility Information Solubility in water: 9 g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone
IUPAC Name barium(2+);dinitrate
Formula Weight 261.34
Percent Purity ≥98.5% (Complexometry)
Grade Pure
Chemical Name or Material Barium nitrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
May intensify fire; oxidizer.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfor
EINECSNumber 233-020-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.