Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%

Catalog No. AA0065318
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0065318 50 g
AA0065306 5 g
AA0065330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA0065318 Supplier Thermo Scientific Chemicals Supplier No. 00065318
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7440-39-3 | Ba | 137.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

Color Silver to Gray
Appearance Silver Gray pieces
Quantity 50 g
Odor Odorless
Form Pieces
Assay Percent Range 99.2% (Metals basis)
Chemical Name or Material Barium
Hazard Category H260-H304
Precautionary Statement P223-P231+P232-P280-P301+P310-P331-P335+P334-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-149-1
RTECSNumber CQ8370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.