Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium rod, 22mm (0.9 in.) dia. x 450mm (17.7 in.), 99+% (metals basis), Sr =0.8%, Thermo Scientific™

Catalog No. AA41302KF
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41302KF 1 Pc.
AA41302KJ 5 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41302KF Supplier Thermo Scientific Chemicals Supplier No. 41302KF

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

Color Gray
Quantity 1 Pc.
Odor Odorless
Weight ≈630g/rod
Form Rod
UN Number UN1400
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) Sr ≤0.8%
Assay (metals basis)
Chemical Name or Material Barium rod, 22mm (0.9 in.) dia. x 450mm (17.7 in.)
Hazard Category Danger
Hazard Statement GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.
Precautionary Statement GHS P Statement
P210-P231+P232-P280-P370+P378i-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: BARIUM
RTECSNumber CQ8370000
TSCA TSCA
storageNote1
  • Packaged under argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.