Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Standard, 1000 μg/mL, 125 mL

Catalog No. USICP056
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-ICP-056 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-ICP-056 Supplier Agilent Technologies Supplier No. ICP056
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-39-3 , 7697-37-2 , 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

CAS Max % 97.9, 2.0, 0.1
Analyte or Component Names Barium,water,nitric acid
Element Nitric Acid, Ba
Solvent or Matrix Water/Dilute Nitric Acid
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Barium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.