Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium thiosulfate, pure, Thermo Scientific™

Catalog No. AC453810025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453810025 2.5 kg
AC453811000 100 g
AC453815000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC453810025 Supplier Thermo Scientific Chemicals Supplier No. 453810025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 35112-53-9
Molecular Formula BaO3S2
Molecular Weight (g/mol) 249.46
InChI Key ONPIOWQPHWNPOQ-UHFFFAOYSA-L
Synonym barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo
PubChem CID 169663
IUPAC Name barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES [O-]S(=O)(=S)[O-].[Ba+2]

Specifications

Quantity 2.5 kg
Linear Formula BaS2O3
Merck Index 15,995
Solubility Information Solubility in water: slightly soluble. Other solubilities: insoluble in alcohol
Formula Weight 249.46
Percent Purity 97.5 to 102.5%
Grade Pure
Chemical Name or Material Barium thiosulfate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 252-375-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.