Learn More
Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide, 99.5+% (metals basis), produces 5% BaTiO3, in isopropanol, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 500 mL |
| UN Number | UN1219 |
| Sensitivity | Moisture Sensitive |
| Formula Weight | 623.89 |
| Percent Purity | ≥99.5% |
| Physical Form | Liquid |
| Assay | (metals basis) |
| Chemical Name or Material | Barium titanium (IV) 2-ethylhexanoate pentaisopropoxide, Produces 5% BaTiO3, in isopropanol |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H225-H361-H302-H332-H336 Highly flammable liquid and vapor. Suspected of damaging fertility or the unborn child. Harmful if swallowed. Harmful if inhaled. May cause drowsiness or dizziness. |
| Precautionary Statement | GHS P Statement P210-P280h-P302+P352 Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/clothing. IF ON SKIN: Wash with plenty of soap and water. |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.