Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium zirconium isopropoxide, 10% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. AA42302AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42302AC 25 mL
AA42302AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42302AC Supplier Thermo Scientific Chemicals Supplier No. 42302AC

Precursor of barium zirconium oxide using Sol-Gel method

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00798551

Specifications

Packaging ChemSeal™ bottles
Quantity 25 mL
UN Number UN3274
Sensitivity Moisture Sensitive
Formula Weight 583.08
Concentration or Composition (by Analyte or Components) 10% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Barium zirconium isopropoxide
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H302-H332-H336
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
storageNote1
  • Packaged under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.