Description
- Designed for testing spatial learning and memory.
- Within the maze, animals are motivated to escape from the brightly lit, open platform to a small dark recessed chamber (escape box).
- During testing, the animal learns the spatial location of the escape box.
- Optional false escape boxes help to remove visual cues that might be observed from a distance or through an open hole.
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Animal | Mouse |
| Includes | False Box |