Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Specifications
Specifications
| Feedwater Source | Deionized, Tap |
| For Use With (Application) | Water purification cartridge |
| For Use With (Equipment) | Nanopure Infinity Systems |
| Includes | One 09-050-226 Tap/DI Pretreatment, three 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters |
| Model | D50256 |
| Type | Cartridge Kit |