Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Feedwater Source | Tap |
| For Use With (Application) | Water purification cartridge |
| For Use With (Equipment) | Nanopure Quad-style System |
| Includes | One 09-034-5C Macropure™ Pretreatment, 09-034 high capacity ion exchange, 09-050-276 Ultrapure™ and 09-050-267 Organicfree cartridges |
| Model | D4802 |
| Type | Cartridge Kit |