Learn More
Thermo Scientific™ Barnstead™ Classic Stills
Description
Remove inorganic solids, organics, bacteria and pyrogens from your water through the distillation process using Thermo Scientific™ Barnstead™ Classic Stills. Choose from bench or floor mounted models. Available in five capacities to meet your production needs, stills are constructed of copper and bronze with a pure tin coating. The inert nature of tin prevents leaching of contaminants into water. Portable stills allow for quick set-up, while floor model electric stills improve safety with over-temperature protection. Steam stills feature optional automatic controls for unattended operation.
- Double-walled boiler and preheating of feed water conserves electricity.
- Vented condenser allows for stripping of gaseous impurities.
- Unique de-concentrator removes scale forming impurities from the boiler.
- Unique Q-Baffle ensures high-quality pyrogen-free water by stripping contaminant laden water droplets from steam.
- Still includes space-saving horizontal condenser.
- Metal construction withstands years of use.
- Inert pure tin pathways assure product water quality.
Portable Stills:
- Require no permanent plumbing or electrical connections allowing for quick setup
- Include low water cutoff for safety protection
- Bench mounted
Floor Model Electric Still:
- Comes standard with floor mounting stand
- Improves safety with new overtemperature protection system
- Allows for easier installation with new plug-and-play hardware
- Processes capacities from 1 to 10 gal./hr.
- Includes low water cutoff that protects heating elements when water supply is interrupted
- Allows for unattended operation with optional fully automatic controls
- Stops still when storage tank is full, starts still when water level falls below preset level
- Drains still boiler at selected two-, four-, eight- and 16-hour intervals to reduce scale formation
Steam Still:
- Utilizes available in-house steam as heat source
- Optional fully automatic controls allow for unattended operation
- Stops still when storage tank is full, starts still when water level falls below preset level
- Drains still boiler at selected two-, four-, eight- and 16-hour intervals to reduce scale formation
- Steam pressure: 35 to 50psi
- Boiler horsepower: 3.5
- Steam flowrate: 90 lb./hr. (41kg/hr.)
Ordering Information:
Automated control systems are available for all stills: G2100 (for 1 gal./hr., 2 gal./hr. models), G2110 (5 gal./hr.), G2125 (10 gal./hr.) and G2010 (10 gal./hr.). Order separately.
This product is not available in all regions.
Includes:
Floor stand (exept for Portable Model A1007).
Requires:
Storage reservoir, UV light assembly and Ventgard Air Filter (order separately)
Recommended for:
Buffer/reagent prep, glassware washing, dishwasher feed
Specifications
Specifications
| Type | Electric |
| Voltage | 208 V |
| Phase | 3 |
| Amperage | 36 A |
| Frequency | 50/60 Hz |
| Output Flow Rate | 5 gal./hr. |
| Dimensions (L x W x H) | 14 x 35 x 77 in. (36 x 89 x 196 cm) |
| Flow Rate | 5 gal./hr. |
| Electrical Requirements | 208 V 50/60 Hz |
| Model | Floor Electric with Stand |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.