Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps

Catalog No. 13998293
Change view
Click to view available options
For Use With (Equipment):
GenPure (UV, UV/UF, UV-TOC and UF/UV-TOC Only), LabTower EDI
MicroPure, Smart2Pure (UV and UV/UVF Only)
Pacific TII; LabTower TII
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-998-293 Pacific TII; LabTower TII
10-451-025 MicroPure, Smart2Pure (UV and UV/UVF Only)
10-451-031 GenPure (UV, UV/UF, UV-TOC and UF/UV-TOC Only), LabTower EDI
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-998-293 Supplier Thermo Scientific™ Supplier No. 50139226
Add to Cart
Add to Cart

Maintain UV function with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps.

Maintain UV function with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps, for use with Barnstead point of use water purification systems.

Ideal for use with the following Barnstead point of use water purification systems:

09.1002:

  • MicroPure™ (UV and UV/UVF systems only)
  • Smart2Pure™ (UV and UV/UVF systems only)

09.2002:

  • GenPure™ (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • GenPure™ Pro (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • GenPure™ xCAD (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • LabTower™ EDI

09.4002:

  • Pacific™ TII
  • LabTower™ TII

09.5002:

For storage reservoirs (230V only) used with the following Barnstead water purification systems:

  • LabTower RO
  • LabTower TII
  • Pacific TII (30 L, 60 L, 100 L only)
  • Smart2Pure (12 only)
TRUSTED_SUSTAINABILITY

Specifications

Description Replacement UV lamp, for use with Pacific TII, LabTower TII
For Use With (Equipment) Pacific TII; LabTower TII
Type UV Lamp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.