Learn More
Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Ultrafilters
Description
Lengthen the life of your water purification system with replacement Thermo Scientific™ Barnstead™ Point-of-Use Water Purification Systems Ultrafilters. For use with Barnstead point-of-use water purification systems, these internal ultrafilters are flushed automatically to allow for a long two-year lifetime.
Use with the following Barnstead point-of-use water purification systems:
Part 50133980:
- GenPure™†
- GenPure™ Pro†
- GenPure™ xCAD†
- Smartu2Pure Pro
†UF, UF/UV and UF/UV-TOC units only
Part 50133981:
- MicroPure™‡
- Smart2 Pure™‡
‡UF and UF/UV units only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Ultrafilter; For use with GenPure, GenPure Pro, GenPure xCAD (UF, UF/UV and UF/UV-TOC units only) |
| For Use With (Equipment) | GenPure, GenPure Pro, GenPure xCAD |
| Type | Ultrafilter |