Learn More
Accuform™ Barricade Tape - KEEP 6 FT DISTANCE
Catalog No. 19169764 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Black, Orange
Black, Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-169-764 Black, Orange
19-169-765 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-169-764 Supplier Accuform™ Supplier No. MPT305
Accuform™ Barricade Tape temporarily sections-off designated areas quickly and effectively as a reminder to practice social and physical distancing to prevent the spread of the Coronavirus (COVID-19)
  • 3-in wide barricade tape on a 1,000-foot roll
  • Flexible, 3-mil polyethylene plastic has printed, repeating message
  • Durable for indoor and outdoor use
  • Easy to cut, wrap, tie, staple, tape, or tie off
  • Meets ANSI Z535.5-3.2 requirements
Specifications
Type Barricade Tape
Thickness 3 mil
Dimensions (L x W) 1000 ft. x 3 in.
Color Black, Orange
Material Polyethylene
Adhesive Type Non-adhesive
Certifications/Compliance ANSI Z535.5-3.2