Learn More
Ansell Guardall Face Shields
Catalog No. p-24316 Shop All Ansell Products
Change view
Click to view available options
Style:
3/4 length
Catalog No. Style
18-999-2885 3/4 length
1 options
This item is not returnable. View return policy
Catalog No. 18-999-2885 Supplier Ansell Supplier No. 41205
This item is not returnable. View return policy
Designed to ensure undistorted visibility and accommodate eyeglass wearers.
  • Full-length face shield reaches from top of head to top of chest
  • Three-quarter length face shield covers from top of head to just below the chin
  • Both styles equipped with a foam band and headband, resistant to fogging
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Includes Foam band and headband, resistant to fogging
Size 3/4 length
Style 3/4 length