Learn More
Thomas Scientific TIP, 1000UL BARRIER TIP(1250UL CAPACITY)STER., 6X96
SDP
Supplier:  Thomas Scientific 1159M42
Encompass
SHARP precision barrier tips eliminate false signals and cross contamination caused by aerosols. SHARP tips feature Denville's new low retention polymer technology that virtually eliminates sample retention. Our inert hydrophobic barrier prevents pipettor contamination without trapping liquid or contaminating samples. These tips provide the most accurate sample delivery in the industry. They contain no nucleic acids, PCR inhibitors, detectable nucleases or endotoxins. All sizes are natural color, presterilized and free of metallic content.
Catalog No. NC1597318
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.