Learn More
Invitrogen™ Barstar (Ribonuclease inhibitor) Polyclonal Antibody

Description
Barstar (Ribonuclease inhibitor) Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | Barstar (Ribonuclease inhibitor) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.70 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene Alias | Barstar |
| Host Species | Rabbit |
| Immunogen | Recombinant Bacillus amyloliquefaciens Barstar (Ribonuclease inhibitor) protein (2-90aa ). |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.