Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix
BIOPHARMAGSA_VA

Catalog No. MT15090CV Shop All Corning Products
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
With Additives:
Glutamine
Glutamine, HEPES
None
Without Additives:
No Glutamine
No Phenol Red
None
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
MT15090CV 6 x 500 mL None No Glutamine
MT10092CV 6 x 500 mL Glutamine, HEPES None
MT16405CV 6 x 500 mL Glutamine No Phenol Red
MT10090CV 6 x 500 mL Glutamine None
MT10092CM 6 x 1 L Glutamine, HEPES None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. MT15090CV Supplier Corning™ Supplier No. 15090CV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for culture of epithelial, endothelial, and granulosa cell types under low serum conditions. Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix has low levels of endotoxin, and absence of contamination.

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Fifty-fifty mixture of DMEM and F-12, originally formulated for rat neuroblastoma cells and MDCK cells
  • Serum-free growth may be possible with appropriate protein supplements
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • This formulation of DMEM/Ham's F-12 is made without L-glutamine
  • Sterile

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM and Ham's F-12, 50/50 Mix
Sterility Sterile
With Additives None
Without Additives No Glutamine
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6 x 500 mL
Endotoxin Level <0.25EU/mL
pH 7 to 7.4
Content And Storage 2° to 8°C
Show More Show Less

IVD Product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.