Learn More
Mopec Base Cabinet, Standing Height, 8 Drawer
SDP
Catalog No. 21381166 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Base Cabinet
Catalog No. Product Type
21-381-166 Base Cabinet
1 options
Catalog No. 21-381-166 Supplier Mopec Supplier No. LC12448S
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • Standing height base cabinet
  • (8) Drawers 7.4 in. H, leveling bolts
  • 4 in. aluminum handles.
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • LC124-48S: 48 in. L x 22 in. W x 35.1 in. H
Specifications
Product Type Base Cabinet
Color Light Gray
Material Stainless Steel