Learn More
Cytiva (Formerly Pall Lab) 47 mm Filter Funnels, Glass - Fritted glass support base with sidearm (1/pkg)#N/A
Catalog No. 502062999 Shop All Cytiva Products
Encompass
The 47 mm glass filter funnels are made of 100% borosilicate glass and they offer excellent chemical compatibility even with the most aggressive solvents.-Borosilicate glass-Chemical compatibility-Ideal for filtration and degassingThe glass filter funnels are ideal for filtration and degassing of HPLC solvents, aqueous mobile phase solutions, and buffers.Ordering information: Materials of Construction: All parts are borosilicate glass except for the clamp and the stopper. The clamp is made of aluminum. The number 8 stopper is made of silicone. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-2999 Supplier Cytiva Supplier No. 4017
The 47 mm glass filter funnels are made of 100% borosilicate glass and they offer excellent chemical compatibility even with the most aggressive solvents.-Borosilicate glass-Chemical compatibility-Ideal for filtration and degassingThe glass filter funnels are ideal for filtration and degassing of HPLC solvents, aqueous mobile phase solutions, and buffers.Ordering information: Materials of Construction: All parts are borosilicate glass except for the clamp and the stopper. The clamp is made of aluminum. The number 8 stopper is made of silicone. This product is not approved for use in medical, clinical, surgical or other patient applications.