Learn More
New Pig Base Neutralizing Absorbent Mat Pad
SDP
Catalog No. 18389134 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
18-389-134 Polypropylene
1 options
This item is not returnable. View return policy
Catalog No. 18-389-134 Supplier New Pig Supplier No. MAT353
Add to Cart
Add to Cart
This item is not returnable. View return policy
Absorb, encapsulate and neutralize small spills of bases in one easy step for safer cleanup. Color indication identifies the best time to handle.
  • Encapsulate and neutralize common bases with these mediumweight pads; great for cleaning up spills in areas where chemicals are stored, mixed or transported
  • Changes color on contact and during neutralization with liquid bases (pH above 7); color change continues until pH becomes neutral and is safe for handling
  • Gelling agents solidify bases during neutralization; prevents them from leaching out of mat
Specifications
Material Polypropylene