Learn More
New Pig Base Neutralizing Absorbent Pillow
SDP
Catalog No. 18389140 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
18-389-140 Polypropylene
1 options
This item is not returnable. View return policy
Catalog No. 18-389-140 Supplier New Pig Supplier No. PIL353
Add to Cart
Add to Cart
This item is not returnable. View return policy
Absorb, encapsulate and neutralize bases in one easy step for safer spill cleanup. Color indication identifies the best time to handle.
  • Encapsulate and neutralize common bases; great for cleaning up spills in areas where chemicals are stored, mixed or transported
  • Changes color on contact and during neutralization with liquid bases (pH above 7); color change continues until pH becomes neutral and is safe for handling
  • Instantly absorbs up to 50% concentrations of potassium hydroxide and other strong chemical bases
  • Gelling agents solidify bases during neutralization; prevents them from leaching out of pillow
Specifications
Material Polypropylene