Learn More
New Pig Base Neutralizing Absorbent Sock
SDP
Catalog No. 18389138 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polymer
Catalog No. Material
18-389-138 Polymer
1 options
This item is not returnable. View return policy
Catalog No. 18-389-138 Supplier New Pig Supplier No. PIG353
Add to Cart
Add to Cart
This item is not returnable. View return policy
Absorb, encapsulate and neutralize bases in one easy step for safer spill cleanup. Color indication identifies the best time to handle.
  • Encapsulate and neutralize the most common base spills; great for battery charging, transport and storage areas
  • Changes color on contact and during neutralization with liquid acids (pH below 7); color change continues until pH becomes neutral and is safe for handling
  • Instantly absorbs up to 98% concentrations of sulfuric acid
  • Gelling agents solidify acids during neutralization; prevents them from leaching out of sock
Specifications
Material Polymer