Learn More
New Pig Base-Neutralizing Spill Kit in Bucket
SDP
Catalog No. 18389122 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Base-Neutralizing Spill Kit in Bucket
Catalog No.
18-389-122 Type: Base-Neutralizing Spill Kit in Bucket
1 options
This item is not returnable. View return policy
Catalog No. 18-389-122 Supplier New Pig Supplier No. KIT319
  • $563.50
    $452.65 / Each of 1
    Save $110.85
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safely respond to corrosive spills with this specially equipped spill kit containing neutralizing absorbents and PPE.
  • All absorbents in this specially-designed kit neutralize base or caustic spills
  • Base Neutralizing Socks stop spreading spills; Base Neutralizing Mats and Pillows soak up puddled liquid
  • Changes color on contact and during neutralization with liquid bases (pH above 7); color change continues until pH becomes neutral and is safe for handling
  • Gelling agents solidify bases during neutralization; prevents them from leaching out
  • Lightweight, 100% polyethylene container resists chemicals
  • Container's molded-in handles make it easy to carry
  • Weather-proof, hinged, polypropylene lid allows easy access and keeps contents clean, dry and secure
  • Contents are organized for quick, efficient response Includes PPE to guard against caustic materials
Specifications
Material Polyethylene