Learn More
Cytiva Sentino Magnetic Filter Funnel Base with support screen for 150 and 300 mL (1/pkg)
Supplier:  Cytiva 4279
Encompass_Preferred
Designed for Use with the New Sentino Microbiology Pump-Variety of sizes - Available in 150, 300, and 500 mL. Convenient 150 mL size easily fits into small autoclaves; 500 mL size is ideal for large sample filtration.-Easy to use - No-leak magnetic seal allows one-handed operation.-Safe to use - Sturdy, polyphenylsulfone plastic construction is shatterproof and offers broad range of chemical compatibility.-Simple placement - Membrane guides help center filter on platform.-Quick point of entry - Forceps access point allows easy filter retrieval.-Accurate - Graduated at 50 mL increments for precise sample measurement. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3024
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.