Learn More
Protective Industrial Products Base35™ 48 in. PVC Raincoat
Catalog No. 19036627 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-036-627 4X-Large
19-036-623 Small
19-036-622 Medium
19-036-621 Large
19-036-624 X-Large
19-036-625 2X-Large
19-036-626 3X-Large
19-036-620 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-036-627 Supplier Protective Industrial Products Supplier No. 4148/XXXXL
Add to Cart
Add to Cart
Base35™ 48 in. PVC Raincoat - Ideal for construction, municipalities, shipyards, agriculture chemical spraying and industrial plant maintenance.
  • Made with 0.35 mm thick PVC and polyester fabric
  • Detachable hood with drawstring
  • Storm flap front with snap closure
  • 2 flapped front pockets with snap closures to secure valuables
  • Cape vented back
  • Corduroy collar for comfort
Specifications
Closure Type Snaps
Color Yellow
Cuff Style Hemmed
Disposable Reusable
Gender Unisex
Material PVC, Polyester
Neck Style Hooded
No. of Pockets 2
Size 4X-Large
Style Duster
Construction Water Resistant, Cape Vented Back, Corduroy Collar
Resistance Tough
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.