Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Basic Botany Models Slide Set
SDPSCI_ED

Catalog No. S89556 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89556 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89556 Supplier United Scientific™ Supplier No. UNSLBOT25
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

United Scientific™ Basic Botany Models Slide Set is a collection of slides specific to the study of botany. Set of 25.

  • Glass prepared microscope slides, supplied in a plastic case.
  • A collection of slides specific to the study of botany.
  • Set of 25 in a plastic case
    • Chlamydomonas, E
    • Spirogyra, vegetative filaments, E
    • Spirogyra, conjugation, E
    • Fucus, dioecious, antheridial and oogonial
    • Mucor, conjugation Conceptacles on one slide, VS
    • Saccharomyces (yeast), budding, E
    • Penicillium, mycelium and conidia, E
    • Marchantia (liverwort), thallus, antheridia, VS
    • Marchantia thallus, archegonia, VS
    • Mnium (moss), antheridial head, VS
    • Marchantia developing sporogonium, VS
    • Mnium, archegonial head, VS
    • Dryopteris (fern), prothallus with antheridia
    • Dryopteris, pinnule with sori, TS and archegonia, E
    • Pinus (pine), leaf, TS
    • Allium (onion), root apex, mitosis, LS
    • Zea, (maize), leaf, TS
    • Helianthus (sunflower), young root, TS
    • Zea, stem, TS
    • Helianthus, stem, TS
    • Cucurbita (marrow), stem LS for sieve tubes
    • Tilia (lime), young and old stem, TS
    • Ranunculus (buttercup), flower bud, LS
    • Prunus (cherry), leaf, TS
    • Pollen grains, wind and insect distribution, E

Specifications

Grade 5 to 12
Format Kit
Product Type Slide Set
Includes Set of 25
Languages English
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.