Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Basic Fuchsin, 88%, pure

Catalog No. AC419140025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419140025 2.5 kg Plastic drum
AC419141000 100 g Glass bottle
AC419145000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419140025 Supplier Thermo Scientific Chemicals Supplier No. 419140025
Add to Cart

CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-99-5
Molecular Formula C20H20ClN3
Molecular Weight (g/mol) 337.85
MDL Number MFCD00012569
InChI Key AXDJCCTWPBKUKL-UHFFFAOYSA-N
Synonym Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride
PubChem CID 12447
IUPAC Name 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Absorbance 1800 (at lambda min.)
Melting Point 250.0°C
CAS Min % 83.0
CAS Max % 100.0
Absorption Ratio 1.1 to 1.4
Dye Content 83% min. (on dry substance)
Assay Percent Range 88%
Beilstein 13, 765
Merck Index 15, 5715
Solubility Information Solubility in water: 4g/L (25°C). Other solubilities: soluble in alcohol and acids, practically insoluble in ether, soluble 30g/L in ethanol (25°C)
Infrared Spectrum Authentic
Lambda Max 549 to 555nm (in 50:50 IMS:water)
Formula Weight 337.84
Percent Purity 88%
Grade Pure
Packaging Plastic drum
Color Green
Quantity 2.5 kg
Flash Point 200°C
Physical Form Fine Powder
Chemical Name or Material Basic Fuchsin, 88%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-189-6
RTECSNumber CX9850000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.