Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basic Green 1 [Ion association reagent for spectrophotometric analysis], TCI America™
SDP

Catalog No. A51061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51061G Supplier TCI America Supplier No. A51061G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ion association reagent for spectrophotometric analysis with 90% minimum purity.

Chemical Identifiers

CAS 633-03-4
Molecular Formula C27H34N2O4S
Molecular Weight (g/mol) 482.639
MDL Number MFCD00011880
InChI Key NNBFNNNWANBMTI-UHFFFAOYSA-M
Synonym Brilliant Green, Diamond Green
PubChem CID 12449
IUPAC Name [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]

Specifications

Color Green
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 482.64
Percent Purity ≥90.0% (Gravimetric Method)
Grade Laboratory
Chemical Name or Material Basic Green 1 [Ion association reagent for spectrophotometric analysis]
EINECSNumber (5)-2031
RTECSNumber BP6825000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.