CAS RN 633-03-4
Brilliant Green (Certified Biological Stain), Fisher Chemical
CAS: 633-03-4 Molecular Formula: C27H34N2O4S Molecular Weight (g/mol): 482.639 MDL Number: MFCD00011880 InChI Key: NNBFNNNWANBMTI-UHFFFAOYSA-M Synonym: Ethyl Green,Basic Green,Malachite Green G PubChem CID: 12449 IUPAC Name: [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
Thermo Scientific Chemicals Brilliant Green, pure, high purity biological stain
CAS: 633-03-4 Molecular Formula: C27H34N2O4S Molecular Weight (g/mol): 482.639 MDL Number: MFCD00011880 InChI Key: NNBFNNNWANBMTI-UHFFFAOYSA-M Synonym: Basic Green 1,C.I. 42040,Ethyl Green PubChem CID: 12449 IUPAC Name: [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
Brilliant Green, ≥85%, Sigma-Aldrich
Dye and antiseptic agent for diagnostic assay manufacturing, hematology, and histology.
Brilliant Green, Sigma-Aldrich
Certified biological stain for microbiological staining, identification, and bacteriostatic use.
Brilliant Green, MP Biomedicals™
CAS: 633-03-4 Molecular Formula: C27H34N2O4S Molecular Weight (g/mol): 482.639 InChI Key: NNBFNNNWANBMTI-UHFFFAOYSA-M Synonym: Astradiamant green GX, Basic Green 1, Diamond Green, Emerald Green, Ethyl Green, Malachite Green G, Solid Green JO PubChem CID: 12449 IUPAC Name: [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
Basic Green 1 [Ion association reagent for spectrophotometric analysis], TCI America™
CAS: 633-03-4 Molecular Formula: C27H34N2O4S Molecular Weight (g/mol): 482.639 MDL Number: MFCD00011880 InChI Key: NNBFNNNWANBMTI-UHFFFAOYSA-M Synonym: Brilliant Green, Diamond Green PubChem CID: 12449 IUPAC Name: [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
Basic Green 1, TCI America™
CAS: 633-03-4 Molecular Formula: C27H34N2O4S Molecular Weight (g/mol): 482.639 MDL Number: MFCD00011880 InChI Key: NNBFNNNWANBMTI-UHFFFAOYSA-M Synonym: Brilliant Green, Diamond Green PubChem CID: 12449 IUPAC Name: [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
Brilliant Green, Certified, CI 42040, Ricca Chemical
CAS: 633-03-4 Molecular Formula: C27H34N2O4S Molecular Weight (g/mol): 482.639 InChI Key: NNBFNNNWANBMTI-UHFFFAOYSA-M PubChem CID: 12449 IUPAC Name: [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
Brilliant Green, 1% (w/v), Ricca Chemical
Certified 1% (w/v) Brilliant Green aqueous solution.
Brilliant Green, 1% Aqueous, pH 0.0 to 2.6 Yellow to Green, Certified, LabChem™
Certified 1% aqueous solution of brilliant green with a pH transition range of 0.0 to 2.6.
Brilliant Green, Reagent Grade, LabChem™
CAS: 633-03-4 Molecular Formula: C27H34N2O4S Molecular Weight (g/mol): 482.639 InChI Key: NNBFNNNWANBMTI-UHFFFAOYSA-M PubChem CID: 12449 IUPAC Name: [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
Thermo Scientific Chemicals Brilliant Green
CAS: 633-03-4 Molecular Formula: C27H34N2O4S Molecular Weight (g/mol): 482.639 MDL Number: MFCD00011880 InChI Key: NNBFNNNWANBMTI-UHFFFAOYSA-M Synonym: Basic Green 1; C.I. 42040 PubChem CID: 12449 IUPAC Name: [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
MilliporeSigma™ Brilliant Green, Hydrogen Sulfate Stain
Stains bacteriological sample material of human origin