Learn More
Eisco™ Basic Magnetic Field Chamber
SCI_ED
Catalog No. S12974 Shop All Eisco Products
Change view
Click to view available options
Grade:
Large
Catalog No. Grade
S12974 Large
1 options
Catalog No. S12974 Supplier Eisco™ Supplier No. PH0805
Add to Cart
Add to Cart
For demonstrations to visualize magnetic fields in three-dimensions.
  • Large magnetic field chamber
  • Three-dimension iron filings field visualizer
  • Iron filings sealed inside chamber
  • Insert bar magnet for three-quarters of an inch in dia. into inner chamber
  • Three-dimensional magnetic field lines appear
Specifications
Description Three-dimension iron filings field visualizer
Grade Large
Product Type Magnetic Field Chamber
Size O.D. x I.D.: 2.31 x 0.62 in.
Dimensions (L x W) O.D. x I.D.: 2.31 x 0.62 in.