Learn More
United Scientific™ Basic Simple Cells Kit
SDPSCI_ED
Catalog No. S139457 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
S139457 Plastic
1 options
This item is not returnable. View return policy
Catalog No. S139457 Supplier United Scientific™ Supplier No. SCLKITE
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quickly and easily demonstrate how a simple cell works with this Basic Simple Cells Kit.
  • This kit allows students to explore the electrical properties of different electrode materials.
  • Demonstrations include voltage, current, and polarization – using safe, everyday electrolytes such as soda, vinegar, and fruit juice.
  • Contents include:
    • A simple cell with electrode holders
    • Electrodes of copper, aluminum, iron, and zinc
    • A mounted resistor for use as a current load
    • Multimeter
    • Manual
Specifications
For Use With (Application) Learn about electrical properties of different electrode materials
Material Plastic
Product Type Simple Cell Kit
Warning: Cancer and reproductive harm
Prop 65