Learn More
Medchemexpress LLC Basroparib | 1858179-75-5 | 98.6% | C18H21F2N7O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14724510MG
Encompass_Preferred
Basroparib is a small-molecule poly(ADP-ribose) polymerase (PARP) inhibitor supplied for research use. It is used in biochemical and cellular studies to probe DNA damage response and PARP-related mechanisms. The compound is provided as a powder with reported purity of 98.64%, molecular formula C18H21F2N7O3, and CAS 1858179-75-5.
  • Selective inhibition of PARP activity for mechanistic studies.
  • Suitable for in vitro biochemical and cellular assays.
  • High reported purity supports analytical and assay reproducibility.
  • Stable powder form for storage and handling convenience.
  • Characterized by molecular formula C18H21F2N7O3.
Catalog No. 50-002-58838
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.