Learn More
Kingfisher Biotech Inc CXCL14 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1784025
Encompass
The Canine, Equine, Human, Monkey, Ferret, Dolphin, Jamaican fruit-eating Bat CXCL14 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine, Equine, Human, Monkey, Ferret, Dolphin, Jamaican fruit-eating Bat CXCL14 applications are for cell culture, ELISA standard, and Western Blot Control. Canine, Equine, Human, Monkey, Ferret, Dolphin, Jamaican fruit-eating Bat CXCL14 yeast-derived recombinant protein can be purchased in multiple sizes. Canine, Equine, Human, Monkey, Ferret, Dolphin, Jamaican fruit-eating Bat CXCL14 Specifications: (Molecular Weight: 9.4 kDa) (Amino Acid Sequence: SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIITTKS VSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE (77)) (Gene ID: n/a). For research use only. Made in the USA
Catalog No. 50-253-2775
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.