Learn More
Kingfisher Biotech Inc Bat IFN Lambda 3 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1828BT025
Encompass_Preferred
The Common Vampire Bat IFN lambda 3 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Common Vampire Bat IFN lambda 3 applications are for cell culture, ELISA standard, and Western Blot Control. Common Vampire Bat IFN lambda 3 yeast-derived recombinant protein can be purchased in multiple sizes. Common Vampire Bat IFN lambda 3 Specifications: (Molecular Weight: 23.4 kDa) (Amino Acid Sequence: PPPLLTLSMP SLVSPSINTD LPVGCTLVLM LMTTVLTRTA AVPVPTPLSA LPGARGCVVA QFKFLAPQDMKAFRRAKDTLEELLLPKNRSCSSRPFPRTRDLRQLQVWERPVALEAELALTLKVLGSIANSTLGDILDQPLHMLRYIHTKLQACVPAQATAGPRPRGHLHHWLHRLQEASKKESTGCLEASVTFNLFRLLTHDLQCVASGGLCI (214)) (Gene ID: 112320026). For research use only. Made in the USA
Catalog No. 50-253-2724
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.