Learn More
Abcam BATCP, Selective HDAC6 Substrate, Solid, 499.5 Da, 2 mg
Description
- Activity: Selective HDAC6 substrate; active in vitro
- Molecular Weight: 499.5 Da
- Purity: >99%
- Form: Solid
- Storage: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | HDAC6 |
| Substrate Properties | Chemical Substrate |
| Substrate | BATCP |
| Shipping Condition | Ambient |
| Buffer | 100% BATCP |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Formulation | 100% BATCP |
| Quantity | 2 mg |
| For Use With (Application) | HDAC6 substrate, In vitro |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.