Learn More
Thermo Scientific™ Bath Fluids for Thermo Scientific Neslab Baths
Use ethylene glycol for low-temperature applications to -30°C, silicone oil for high temperature applications to +150°C.
$520.50 - $930.00
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
13-265-154
![]() |
Thermo Scientific™
610000000000 |
Silicone Oil |
Each for $930.00
|
|
|||||
13-265-155
![]() |
Thermo Scientific™
610000000001 |
Ethylene Glycol Bath Fluid |
Each for $520.50
|
|
|||||
Description
Use ethylene glycol for low-temperature applications to -30°C, silicone oil for high temperature applications to +150°C.
Use ethylene glycol for low-temperature applications to -30°C, silicone oil for high temperature applications to +150°C.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
In 5 gal. (18.9L) containers.
Specifications
Neslab bath |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.