Learn More
Thomas Scientific Poly Pro Bath (Dual Chamber) - 5.5L capacity (each chamber), 100-240VAC
SDP
Supplier:  Thomas Scientific 1203T12
Encompass
At literally half the cost of other same-sized water baths, the Poly Pro bath has a unique polypropylene chamber. Whether the Poly Pro bath is used to simply keep media warm, or for an experiment that requires precise temperature control this water bath is ideal for all general-purpose laboratory applications. Due to the distinctive nature of polypropylene, not only is the chamber's surface resistant to corrosion and chemical attack, but glossy finish is non-stick for easy clean-up and removal of mineral deposits. Equipped with a low water detection system the Poly Pro will sound an alarm and shut off power to the heater if the water level is below one inch. Features Clear instructions printed on front label The PB-200 is half the cost of other similar sized water baths that offer the same capabilities The durable, nonconductive polypropylene interior and exterior provide 'double insulation' Resistant to virtually all types of chemical attack Patented WaveOn sensing technol
Catalog No. 50-145-7687
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.