Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Batimastat DMSO Solution, MMP Inhibitor, 1 mg

Catalog No. 502704547
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4547 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4547 Supplier Abcam Supplier No. AB1466191MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, broad-spectrum matrix metalloprotease inhibitor with demonstrated anticancer activity in vivo.

  • Potent, broad-spectrum Matrix metalloprotease (MMP) inhibitor
  • IC50 values: 3 nM (MMP-1), 4 nM (MMP-2), 4 nM (MMP-9), 6 nM (MMP-7), and 20 nM (MMP-3)
  • Molecular Weight: 477.6 Da
  • Purity: >98%
  • Anticancer activity in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Matrix Metalloproteases
Shipping Condition Ambient
Buffer 99.56% dimethylsulfoxide
Content And Storage Store at -20°C.
Form Liquid
Formulation 99.56% Dimethylsulfoxide, 0.44% (2S,3R)-N-hydroxy-N'-[(2S)-1-(methylamino)-1-oxo-3-phenylpropan-2-yl]-3-(2-methylpropyl)-2-(thiophen-2-ylsulfanylmethyl)butanediamide
Quantity 1 mg
For Use With (Application) Inhibition of Matrix metalloprotease, Anticancer research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.