Learn More
Siemens Healthineers Batroxobin Reagent
Catalog No. 23044694 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
2 x 5 mL
Catalog No. Quantity
23-044-694 2 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-694 Supplier Siemens Healthineers Supplier No. 10446463
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of batroxobin time
Batroxobin reagent is a snake venom-based reagent intended for the determination of the batroxobin time. It is ideal for monitoring fibrinolytic therapy by determination of fibrinogen/fibrin degradation products, diagnosis of afibrinogenemia and dysfibrinogenemia, and elucidation of prolonged thrombin times in cases of suspected presence of heparin.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Thrombin/Batroxobin Time Testing
Quantity 2 x 5 mL