Learn More
Grainger Battery, Lithium, Size CR2, 3VDC
Supplier:  Grainger 45EK20
Encompass_Preferred
CR2 batteries are often used in digital cameras and photographic equipment. They have a nominal diameter of 16 mm. Disposable specialty batteries provide increased capacity to handle spikes in power demand. They're used in high-drain devices such as digital cameras, Wi-Fi-enabled equipment, tactical flashlights, occupancy sensors, defibrillators, microphones, and faucet sensors. These batteries are typically cylindrical and look similar to standard disposable batteries while providing more power for demanding applications. They can be discarded when their charge runs out.
Catalog No. NC1227968
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.