Learn More
Med Vet International Diagnostic Battery Pack Philips Lithium For Heartstart Frx /Onsite / Hs1/ Home Defibrillator, M5070A, Each
Supplier:  Med Vet International 863134EA
Encompass_Preferred
Non-rechargeable The battery typically lasts 4 years installed keeping your Philips AED in a "ready to use" condition as well as running mission critical self tests The battery has the capacity to deliver a minimum of 200 shocks or 4 hours of patient monitoring5 year shelf life (new and uninstalled). The battery expected expiration date is 4 years from the date you install your battery as long as you install the battery within the "install by date"12V
Catalog No. 50-283-0921