Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Battery Holder Set
SDPSCI_ED

Catalog No. S139487 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139487 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139487 Supplier United Scientific™ Supplier No. BHS001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Set for use in many electrical experiments requiring batteries.

This battery holder fits four D cell batteries (not included) and is useful for virtually any experiment requiring a low voltage power supply and ships fully assembled.

  • Five color-coded binding posts on the base make for easy, safe, and reliable connections to hook-up cords providing outlets for 1.5 V, 3 V, 4.5 V, or 6 V supply voltages
  • Battery holder is firmly mounted on a heavy duty base

Specifications

Grade 5 to 12
Format Kit
For Use With (Equipment) D Batteries
Product Type Battery Holder
Material Plastic
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.