Learn More
Eisco™ Battery Operated LED Ray Box
SCI_ED
Catalog No. S15124 Shop All Eisco Products
Change view
Click to view available options
LED Ray Box
Catalog No.
S15124 LED Ray Box
1 options
Catalog No. S15124 Supplier Eisco™ Supplier No. PH0602ALED
Add to Cart
Add to Cart
Battery Operated LED Ray Box is ideal for classroom demonstrations in physics, optics, reflection, refraction, and more.
  • Battery Operated LED Light Ray Box with 7 Grooves
  • Runs substantially cooler than traditional incandescent lamps
  • Uses the latest, cool ultra bright LED technology giving superior white light
  • Rugged ABS injection molded enclosure
  • Includes two slit plates and one lens
  • Batteries not included
Specifications
Product Type LED Ray Box
Description Battery Operated
Material ABS Plastic
For Use With (Application) Classroom Demonstrations
Includes Lens, 2 Slit Plates, Battery Power Source
Dimensions (L x W x H) 6.25 x 3.5 x 3.25 in.