Description
- Features pistol grip with trigger button.
- 10050 mAh battery provides up to 18 hours of operation.
- Direct backup or replacement for HH85 or EXA81 devices; use with HH83 to add pistol grip with trigger functionality and expand battery life.
- Battery can he charged as stand-alone unit in charging dock (not included) or when installed in EXA81 and HH-series devices.
Specifications
Specifications
| For Use With (Equipment) | HH83 Handheld Barcode Scanner and Mobile Computer (B-HH83-2D), HH83 Handheld RFID Reader (B-HH83-UHF-US),HH85 Handheld RFID Reader with Pistol Grip (B-HH85-UHF-US), Brady EXA81 RFID Reader (B-EXA81-US) |
| Type | Battery Charger |
Has lithium-ion battery - must ship ground