Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. ms711r7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS711R7 7 mL Unconjugated Prediluted Prediluted
MS711P0 100 μL Unconjugated Concentrated 200μg/mL
MS711P 1 mL Unconjugated Concentrated 200μg/mL
MS711P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS711B0 100 μL Biotin Concentrated 200μg/mL
MS711R1 1 mL Unconjugated Prediluted Prediluted
MS711B1 500 μL Biotin Concentrated 200μg/mL
MS711B 1 mL Biotin Concentrated 200μg/mL
MS711P1 500 μL Unconjugated Concentrated 200μg/mL
MS711PABX 200 μL Unconjugated Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS711R7 Supplier Epredia Supplier No. MS711R7

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.

Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).

Host Species: Mouse

Clone: 2D2

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: aa 3-16

Immunogen: A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human Bax protein. This amino acid sequence is not shared by mouse and rat Bax protein.

Molecular Weight: 21kDa

Positive Control: HeLa cells, Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry (Not tested)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Bax Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2D2
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein.
Quantity 7 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.