Learn More
McKesson General Medical BAXTER 2C6572 48/CS
Supplier:  McKesson General Medical 312080
Encompass
CONTINU-FLO Solution Set, Non-Vented, 0.2 Micron Filter, 2 INTERLINK Injection Sites, Backcheck Valve, 60 drops/mL, 105" (2.7 m)
Catalog No. NC1540354
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.