Learn More
Hanna Pharmaceutical Supply Co., Inc. BAXTER CONTINUFLO SOLUTION SET
Supplier:  Hanna Pharmaceutical Supply Co., Inc. 2C6571
Encompass_Preferred
Due to recent natural disasters, production of sterile water has been negatively impacted.  Please expect long lead times on these itemsBaxter Continu-Flo Solution Set, Non-Vented, 0.2um filter, 2 INTERLINK Injection Sites, Backcheck Valve, 10 drops/mL, 105" (2.7 m)
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
Catalog No. NC2020311
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.