Learn More
Selleck Chemical LLC Bay 11-7085 10mg 196309-76-9 Bay 11-7083
SDP
Supplier:  Selleck Chemical LLC S735210MG
Encompass_Preferred
BAY 11-7085 (Bay 11-7083) is an irreversible inhibitor of TNF?-induced I?B? phosphorylation with IC50 of 10 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4674
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.